missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carbon Steel, AISI 1060, G10600, 100g, ARMI∣MBH
Click to view available options
Quantity:
100 g
Unit Size:
100g
Description
Carbon Steel, AISI 1060, G10600, 100g
Specifications
Specifications
| Chemical Name or Material | Boron/Calcium/Niobium/Titanium/Aluminium/Oxygen/Tungsten/Arsenic/Cobalt/Tin/Nitrogen/Phosphorus/Molybdenum/Vanadium/Nickel/Chromium/Copper/Silicon/Carbon/Manganese/Magnesium/Lead/Antimony/Zinc/Zirconium |
| CAS | 7439-89-6 |
| Quantity | 100 g |
| Form | Pieces |
| Concentration or Composition (by Analyte or Components) | Boron 0.0003 %/Calcium 0.0005 %/Niobium 0.001 %/Titanium 0.0017 %/Aluminium 0.002 %/Oxygen 0.002 %/Tungsten 0.002 %/Arsenic 0.0046 %/Cobalt 0.005 %/Tin 0.0069 %/Nitrogen 0.0088 %/Phosphorus 0.0123 %/Molybdenum 0.0176 %/Vanadium 0.023 %/Nickel 0.048 %/Chromium 0.096 %/Copper 0.107 %/Silicon 0.22 %/Carbon 0.63 %/Manganese 0.7 %/Magnesium (0.0004) %/Lead (0.001) %/Antimony (0.002) %/Zinc (0.003) %/Zirconium (0.003) % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction