missing translation for 'onlineSavingsMsg'
Learn More

DL-Asparagine Monohydrate, TRC

Product Code. 30347449
Click to view available options
Quantity:
100 g
Unit Size:
100g
This item is not returnable. View return policy

Product Code. 30347449

Brand: Toronto Research Chemicals TRCA788985100G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material DL-Asparagine hydrate
CAS 69833-18-7
Molecular Formula C4 H8 N2 O3 . H2 O
Quantity 100 g
Synonym DL-Asparagine hydrate, Asparagine, hydrate (1:1), Asparagine, monohydrate (9CI), DL-Asparagine, monohydrate
InChI Formula InChI=1S/C4H8N2O3.H2O/c5-2(4(8)9)1-3(6)7;/h2H,1,5H2,(H2,6,7)(H,8,9);1H2
SMILES O.NC(CC(=O)N)C(=O)O
IUPAC Name 2,4-diamino-4-oxobutanoic acid;hydrate
Molecular Weight (g/mol) 150.13
Formula Weight 150.0641
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.