missing translation for 'onlineSavingsMsg'
Learn More

Benzo[e]pyrene, TRC

Product Code. 30338853
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
This item is not returnable. View return policy

Product Code. 30338853

Brand: Toronto Research Chemicals TRCB205805100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Benzo(e)pyrene
CAS 192-97-2
Molecular Formula C20 H12
Quantity 100 mg
Synonym Benzo[e]pyrene, 1,2-Benzopyrene, 1,2-Benzpyrene, 4,5-Benzopyrene, 4,5-Benzpyrene, NSC 89273
InChI Formula InChI=1S/C20H12/c1-2-8-16-15(7-1)17-9-3-5-13-11-12-14-6-4-10-18(16)20(14)19(13)17/h1-12H
SMILES c1ccc2c(c1)c3cccc4ccc5cccc2c5c34
IUPAC Name benzo[e]pyrene
Molecular Weight (g/mol) 252.31
Formula Weight 252.0939
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.