missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzyldimethyltetradecylammonium-d7 Chloride, TRC
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Specifications
Specifications
| Chemical Name or Material | Benzyldimethyltetradecylammonium-d7 Chloride |
| CAS | 1219178-72-9 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C23H35D7ClN |
| Quantity | 100 mg |
| Synonym | N,N-Dimethyl-N-tetradecylbenzenemethanaminium-d7 Chloride, (Benzyl-d7)dimethyltetradecylammonium Chloride, Arquad DM 14B90-d7, Cation M 2-100R-d7, Cyncal-d7, Dibactol-d7, Myristalkonium-d7 Chloride, Zeph-d7, Zephiramine-d7, Zephiramine-d7 Chloride |
| InChI Formula | InChI=1S/C23H42N.ClH/c1-4-5-6-7-8-9-10-11-12-13-14-18-21-24(2,3)22-23-19-16-15-17-20-23;/h15-17,19-20H,4-14,18,21-22H2,1-3H3;1H/q+1;/p-1/i15D,16D,17D,19D,20D,22D2; |
| SMILES | CCCCCCCCCCCCCC[N+](C)(C)CC1=CC=CC=C1.[Cl-] |
| IUPAC Name | [dideuterio-(2,3,4,5,6-pentadeuteriophenyl)methyl]-dimethyl-tetradecylazanium;chloride |
| Molecular Weight (g/mol) | 375.08 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction