missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carazolol Hydrochloride Salt, TRC
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Specifications
Specifications
| Chemical Name or Material | Carazolol Hydrochloride Salt |
| CAS | 51997-43-4 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C18 H22 N2 O2 . Cl H |
| Quantity | 100 mg |
| Synonym | 2-Propanol, 1-(9H-carbazol-4-yloxy)-3-[(1-methylethyl)amino]-, hydrochloride (1:1) (ACI), 2-Propanol, 1-(9H-carbazol-4-yloxy)-3-[(1-methylethyl)amino]-, monohydrochloride (9CI), 1-((9H-Carbazol-4-yl)oxy)-3-(isopropylamino)propan-2-ol hydrochloride, Carazolol hydrochloride, NSC 305336, 1-(9H-Carbazol-4-yloxy)-3-[(1-methylethyl)amino]-2-propanol monohydrochloride |
| InChI Formula | InChI=1S/C18H22N2O2.ClH/c1-12(2)19-10-13(21)11-22-17-9-5-8-16-18(17)14-6-3-4-7-15(14)20-16;/h3-9,12-13,19-21H,10-11H2,1-2H3;1H |
| SMILES | Cl.CC(C)NCC(O)COc1cccc2[nH]c3ccccc3c12 |
| IUPAC Name | 1-(9H-carbazol-4-yloxy)-3-(propan-2-ylamino)propan-2-ol;hydrochloride |
| Molecular Weight (g/mol) | 334.84 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction