missing translation for 'onlineSavingsMsg'
Learn More

Clemastine fumarate, MedChemExpress

Product Code. 16438226
Encompass_Preferred
Click to view available options
Quantity:
100 mg
200 mg
500 mg
Unit Size:
100mg
200mg
500mg
This item is not returnable. View return policy

Product Code. 16438226

Brand: Medchem Express HYB0298A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Clemastine (HS-592) fumarate is a selective histamine H1 receptor antagonist. Clemastine fumarate is an antihistamine mainly used for relieving symptoms of allergic reactions primarily by competing with histamine to bind H1 receptors. Anti-inflammatory effects.

Clemastine (HS-592) fumarate is a selective histamine H1 receptor antagonist. Clemastine fumarate is an antihistamine mainly used for relieving symptoms of allergic reactions primarily by competing with histamine to bind H1 receptors. Anti-inflammatory effects.

Specifications

Chemical Name or Material Clemastine fumarate
CAS 14976-57-9
Color White
Purity Grade Notes Research
Molecular Formula C25H30ClNO5
Quantity 100 mg
Synonym HS-592 fumarateMeclastine fumarate
Solubility Information DMSO : 14.29 mg/mL (31.07 mM; Need ultrasonic) ∣H2O : 0.67 mg/mL (1.46 mM; Need ultrasonic)
SMILES CN1[C@@H](CCO[C@](C2=CC=CC=C2)(C3=CC=C(Cl)C=C3)C)CCC1.O=C(O)/C=C/C(O)=O
Molecular Weight (g/mol) 459.96
Formula Weight 459.96
Percent Purity 99.41%
Grade Research
Physical Form Powder
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.