missing translation for 'onlineSavingsMsg'
Learn More

L-Asparagine-d3 Hydrate, TRC

Product Code. 30338889
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 30338889

Brand: Toronto Research Chemicals TRCA790007100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material L-Asparagine-d3 Hydrate
CAS 2483831-59-8
Purity Grade Notes HPLC
Molecular Formula C4 D3 H5 N2 O3 . H2 O
Quantity 100 mg
Synonym L-Asparagine-2,3,3-d3 hydrate (1:1), L-Asparagine-2,3,3-D3 monohydrate, (2S)-2,4-Diamino-4-oxobutanoic-2,3,3-d4 acid monohydrate, (S)-2,4-Diamino-4-oxobutanoic acid-d3 monohydrate
InChI Formula InChI=1S/C4H8N2O3.H2O/c5-2(4(8)9)1-3(6)7;/h2H,1,5H2,(H2,6,7)(H,8,9);1H2/t2-;/m0./s1/i1D2,2D;
SMILES O.[2H]C([2H])(C(=O)N)[C@]([2H])(N)C(=O)O
IUPAC Name (2S)-2,4-diamino-2,3,3-trideuterio-4-oxobutanoic acid;hydrate
Molecular Weight (g/mol) 153.1517
Formula Weight 153.0829
Percent Purity >95
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.