missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acrylonitrile(stabilized with 35-45 ppm 4-hydroxy anisole (H750015)), TRC
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
Specifications
Specifications
| Chemical Name or Material | Acrylonitrile |
| CAS | 107-13-1 |
| Purity Grade Notes | GC |
| Molecular Formula | C3 H3 N |
| Quantity | 100 mL |
| Synonym | Acrylonitrile (8CI), Acrylon, Carbacryl, Cyanoethene, Cyanoethylene, Fumigrain, NSC 6362, Propenenitrile, VCN, Ventox, Vinyl cyanide |
| InChI Formula | InChI=1S/C3H3N/c1-2-3-4/h2H,1H2 |
| SMILES | C=CC#N |
| IUPAC Name | prop-2-enenitrile |
| Molecular Weight (g/mol) | 53.06 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction