missing translation for 'onlineSavingsMsg'
Learn More

Enzo Life Sciences 100ml RIPA cell lysis buffer 3, (100 ml)

Brand:  Enzo Life Sciences ADI-80-1496

Encompass_Preferred

100ml RIPA cell lysis buffer 3, (100 ml)

Product Code. 15145507

  • 284.35 € / 100mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • RIPA cell lysis buffer 3, (100 ml)

Signal Word
  • Warning

Hazard Category
  • Respiratory sensitiser Category 1

Hazard Statement
  • H317-May cause an allergic skin reaction.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P272-Contaminated work clothing should not be allowed out of the workplace.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P321-Specific treatment (see on this label).
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P363-Wash contaminated clothing before reuse.
  • P501-Dispose of contents/container to .

SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.