missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Atlantic Blue™ Mouse IgG2a isotype control (11A1B2), Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech AB-66360-2-100UG
This item is not returnable.
View return policy
Description
The IgG2a isotype control antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of IgG2a isotype control. This antibody recognizes megathura crenulata antigen. The IgG2a isotype control antibody has been validated for the following applications: FC analysis. This antibody is atlantic blue™.Specifications
| IgG2a isotype control | |
| Monoclonal | |
| 200 μg/mL | |
| Mouse | |
| Protein A purification | |
| RUO | |
| M. crenulata | |
| Flow Antibody | |
| IgG2a |
| Flow Cytometry | |
| 11A1B2 | |
| Atlantic Blue | |
| Recombinant Protein | |
| 100 μg | |
| Primary | |
| Store at 2-8°C. Avoid exposure to light. Stable for one year after shipment. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction