missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetyl-d3 Chloride, CDN
Click to view available options
Quantity:
10 g
Unit Size:
10g
Specifications
Specifications
| Chemical Name or Material | Acetyl-d3 Chloride |
| CAS | 19259-90-6 |
| Molecular Formula | C2 2H3 Cl O |
| Quantity | 10 g |
| InChI Formula | InChI=1 S/C2H3ClO/c1-2(3)4/h1H3/i1D3 |
| SMILES | [2 H]C([2 H])([2 H])C(=O)Cl |
| IUPAC Name | 2,2,2-trideuterioacetyl chloride |
| Molecular Weight (g/mol) | 81.52 |
| Formula Weight | 81.0061 g/mol |
| Percent Purity | 99 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction