missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bromoethane-d5, TRC
Click to view available options
Quantity:
10 g
Unit Size:
10g
Specifications
Specifications
| Chemical Name or Material | Bromoethane-D5 |
| CAS | 3675-63-6 |
| Purity Grade Notes | GC |
| Molecular Formula | C2 2H5 Br |
| Quantity | 10 g |
| Synonym | Ethane-1,1,1,2,2-d5, 2-bromo- (ACI), 2-Bromoethane-1,1,1,2,2-d5 (ACI), Ethane-d5, bromo- (6CI, 7CI, 8CI, 9CI), 1-Bromo-1,1,2,2,2-pentadeuterioethane, Bromoethane-d5, Ethyl bromide-d5, Ethyl-d5 bromide, Pentadeuterioethyl bromide, Pentadeuteroethyl bromide, Perdeuterioethyl bromide, Bromoethane-D5 |
| InChI Formula | InChI=1S/C2H5Br/c1-2-3/h2H2,1H3/i1D3,2D2 |
| SMILES | [2H]C([2H])([2H])C([2H])([2H])Br |
| IUPAC Name | 1-bromo-1,1,2,2,2-pentadeuterioethane |
| Molecular Weight (g/mol) | 114 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction