missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium Fluoride, TRC
Click to view available options
Quantity:
10 g
Unit Size:
10g
Specifications
Specifications
| Chemical Name or Material | Calcium Fluoride |
| CAS | 7789-75-5 |
| Molecular Formula | CaF2 |
| Quantity | 10 g |
| Synonym | Calcium fluoride (CaF2), Calcium fluoride (8CI), Calcarea fluorica, Calcium difluoride, Calcium difluoride (CaF2), Fluorite Powder Calcium Fluoride, HO 100, HO 100W, Irtran 3 |
| InChI Formula | InChI=1S/Ca.2FH/h;2*1H/q+2;;/p-2 |
| SMILES | F[Ca]F |
| IUPAC Name | difluorocalcium |
| Molecular Weight (g/mol) | 78.08 |
| Formula Weight | 77.9594 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction