missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p-Toluenesulfonylhydrazide-N,N,N-d3, CDN
Click to view available options
Quantity:
5 g
10 g
Unit Size:
10g
5g
Specifications
Specifications
| Chemical Name or Material | p-Toluenesulfonyl Hydrazide-D3 |
| CAS | 109333-73-5 |
| Molecular Formula | C7 D3 H7 N2 O2 S |
| Quantity | 10 g |
| Synonym | Benzenesulfonic acid, 4-methyl-, hydrazide-1,2,2-d3 (ACI), Benzenesulfonic acid, 4-methyl-, hydrazide-d3 (9 CI), p-Toluenesulfonylhydrazide-N,N,N-d3, 4-Methylbenzenesulfonic acid hydrazide-1,2,2-d3 |
| InChI Formula | InChI=1 S/C7H10N2O2S/c1-6-2-4-7(5-3-6)12(10,11)9-8/h2-5,9 H,8H2,1H3/i/hD3 |
| SMILES | [2 H]N([2 H])N([2 H])S(=O)(=O)c1ccc(C)cc1 |
| IUPAC Name | N,N',N'-trideuterio-4-methylbenzenesulfonohydrazide |
| Molecular Weight (g/mol) | 189.25 |
| Formula Weight | 189.0651 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction