missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-3-methylselenyl Propionic Acid, TRC

Product Code. 30336521
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
This item is not returnable. View return policy

Product Code. 30336521

Brand: Toronto Research Chemicals TRCA63443810MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Amino-3-methylselenyl Propionic Acid
CAS 26046-90-2
Molecular Formula C4 H9 N O2 Se
Quantity 10 mg
Synonym 2-Amino-3-methylselenyl Propionic Acid
InChI Formula InChI=1S/C4H9NO2Se/c1-8-2-3(5)4(6)7/h3H,2,5H2,1H3,(H,6,7)/t3-/m0/s1
SMILES C[Se]C[C@H](N)C(=O)O
IUPAC Name (2R)-2-amino-3-methylselanylpropanoic acid
Molecular Weight (g/mol) 182.08
Formula Weight 182.98
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Tack! Din feedback har skickats. Fisher Scientific arbetar alltid för att förbättra vårt innehåll åt dig. Vi uppskattar din feedback.