missing translation for 'onlineSavingsMsg'
Learn More

Bis(2-methoxyethyl) Phthalate-3,4,5,6-d4, TRC

Product Code. 30341002
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
This item is not returnable. View return policy

Product Code. 30341002

Brand: Toronto Research Chemicals TRCB48751110MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Phthalic acid, bis-methylglycol ester D4
CAS 1398065-54-7
Molecular Formula C14 2H4 H14 O6
Quantity 10 mg
Synonym Phthalic Acid, Bis-methylglycol Ester D4, Phthalic Acid, Bis-methylglycol Ester D4 (phenyl D4)
InChI Formula InChI=1S/C14H18O6/c1-17-7-9-19-13(15)11-5-3-4-6-12(11)14(16)20-10-8-18-2/h3-6H,7-10H2,1-2H3/i3D,4D,5D,6D
SMILES [2H]c1c([2H])c([2H])c(C(=O)OCCOC)c(C(=O)OCCOC)c1[2H]
IUPAC Name bis(2-methoxyethyl) 3,4,5,6-tetradeuteriobenzene-1,2-dicarboxylate
Molecular Weight (g/mol) 286.31
Formula Weight 286.1354
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.