missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bisphenol A-d14, TRC
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Specifications
Specifications
| Chemical Name or Material | Bisphenol A D14 |
| CAS | 120155-79-5 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C15 2H14 H2 O2 |
| Quantity | 10 mg |
| Synonym | Phen-2,3,5,6-d4-ol, 4,4'-[1-(methyl-d3)ethylidene-2,2,2-d3]bis-, 4,4'-[1-(Methyl-d3)ethylidene-2,2,2-d3]bis[phen-2,3,5,6-d4-ol], Bisphenol A-d14, Bisphenol A D14, Bisphenol A (ring,dimethyl-D14), 2,2-Bis(4-hydroxy-2,3,5,6-tetradeuteriophenyl)-1,1,1,3,3,3-hexadeuteriopropane, 4,4'-Isopropylidenediphenol (ring,dimethyl-D14) |
| InChI Formula | InChI=1S/C15H16O2/c1-15(2,11-3-7-13(16)8-4-11)12-5-9-14(17)10-6-12/h3-10,16-17H,1-2H3/i1D3,2D3,3D,4D,5D,6D,7D,8D,9D,10D |
| SMILES | [2H]c1c([2H])c(c([2H])c([2H])c1O)C(c2c([2H])c([2H])c(O)c([2H])c2[2H])(C([2H])([2H])[2H])C([2H])([2H])[2H] |
| IUPAC Name | 2,3,5,6-tetradeuterio-4-[1,1,1,3,3,3-hexadeuterio-2-(2,3,5,6-tetradeuterio-4-hydroxyphenyl)propan-2-yl]phenol |
| Molecular Weight (g/mol) | 242.37 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction