missing translation for 'onlineSavingsMsg'
Learn More
Learn More
O-(2-Acetamido-2-deoxy-D-glucopyranosylidene)amino N-Phenylcarbamate, TRC
missing translation for 'orderingAttributeHoverText'
Quantity:
10 mg
missing translation for 'unitSize'
10mg
Specifications
Specifications
| Chemical Name or Material | O-(2-Acetamido-2-deoxy-D-glucopyranosylidene)amino N-Phenylcarbamate |
| CAS | 132489-69-1 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C15 H19 N3 O7 |
| Quantity | 10 mg |
| InChI Formula | InChI=1S/C15H19N3O7/c1-8(20)16-11-13(22)12(21)10(7-19)24-14(11)18-25-15(23)17-9-5-3-2-4-6-9/h2-6,10-13,19,21-22H,7H2,1H3,(H,16,20)(H,17,23)/b18-14-/t10-,11-,12-,13-/m1/s1 |
| SMILES | CC(=O)N[C@@H]1[C@@H](O)[C@H](O)[C@@H](CO)O/C/1=N\OC(=O)Nc2ccccc2 |
| IUPAC Name | [(Z)-[(3R,4R,5S,6R)-3-acetamido-4,5-dihydroxy-6-(hydroxymethyl)oxan-2-ylidene]amino] N-phenylcarbamate |
| Molecular Weight (g/mol) | 353.33 |
| Formula Weight | 353.1223 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction