Learn More
DiscoverX Phenylephrine
Product Code. 17372479
Encompass
Click to view available options
:
10 mg
Product Code. 17372479
Brand: DiscoverX 921155
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Phenylephrine is an agonist for ADRA1A, B and C. Provided as 10mg of dry powder.
Phenylephrine hydrochloride, or NeoSynephrine, is a synthetic sympathomimetic drug compound structurally similar to epinephrine that is a selective agonist of the α1-adrenergic receptor on blood vessels, causing vasoconstriciton. Its in-vivo effects include decongestion, hypertension, and pupil dilation. It is a common ingredient in many decongestants, and for cold, flu, allergy, or breathing illnesses. It acts as an agonist of ADRA1A, B, and C. Provided as 10mg of dry powder.
Specifications
Quantity 10 mg