Learn More
DiscoverX Serotonin 5-HT
Product Code. 17362439
Encompass
Click to view available options
:
10 mg
Product Code. 17362439
Brand: DiscoverX 921108
Serotonin 5-HT is an agonist for the serotonin (HRT) receptors. Provided as 10mg of dry powder alongside reconstitution buffer.
Serotonin, or 5-hydroxytryptamine (5-HT), is a mono-amine neurotransmitter derived from tryptophan that is responsible for regulation of mood and associated with depression. An agonist for the serotonin (HRT) receptors, it is primarily located in the CNS and GI tract, and modulates physiological functions related to neurotransmission, GI motility, pain perception, hemostasis, hunger, and cardiovascular integrity. Provided as 10mg of dry powder alongside reconstitution buffer.
Specifications
Quantity 10 mg