missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Biosciences™ SU 5402
Click to view available options
:
10 mg
Description
SU 5402 is a potent and selective vascular endothelial growth factor receptor (VEGFR) and fibroblast growth factor receptor (FGFR) inhibitor (IC50 values are 0.02, 0.03, 0.51 and > 100μM at VEGFR2, FGFR1, PDGFRβ and EGFR respectively). Inhibits embryonic left-right determination and exhibits potent anticancer activity in vitro and in vivo. Also attenuates integrin β4-induced differentiation of neural stem cells. Supports mESC self-renewal.
For more information about how SU 5402 may be used, see our protocol: Accelerated Induction of Cortical Neurons from hiPSCs
Specifications
Specifications
| Chemical Name or Material | 2-[(1,2-Dihydro-2-oxo-3H-indol-3-ylidene)methyl]-4-methyl-1H-pyrrole-3-propanoic acid |
| CAS | 215543-92-3 |
| Quantity | 10 mg |
| Target | FGF Receptor Inhibitors |
| Molecular Formula | C17H16N2O3 |
| Purity | 0.95 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction