Learn More
Viking Optical™ 10X Folding Magnifier
Product Code. 12886325
Encompass_Preferred
Change view
Click to view available options
Diameter (Metric) Lens:
11 mm
19 mm
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Lens Unit Size
12886325 19 mm Each
12896325 11 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 12886325 Supplier Viking Optical™ Supplier No. 5609
Estimated Shipment: 21-10-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Pocket doublet type, giving tolerably flat field with minimum chromatic aberration. Lens approximately 19mm diameter. Mounted in metal frame.
Specifications
Diameter (Metric) Lens 19 mm