missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ 10X Strep-Diluent

Product Code. 30000907
Change view
Click to view available options
Quantity:
2 mL
Unit Size:
2mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30000907 2 mL 2mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30000907 Supplier R&D Systems™ Supplier No. 467109604
  • ONLINE EXCLUSIVE PRICE
    111.51 €
    98.70 € / 2mL
    Save 12.81 €
    11% Off
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

R&D Systems™ 10X Strep-Diluent is a solution that is used as a diluent for the Strep-HRP.

  • Hazard Statements (GHS-US):
    • H319 - Causes serious eye irritation
  • Precautionary Statements (GHS-US):
    • P264 - Wash hands, forearms, and other exposed areas thoroughly after handling.
    • P280 - Wear protective gloves, protective clothing, and eye protection.
    • P305+P351+P338 - IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    • P337+P313 - If eye irritation persists: Get medical advice/attention.
  • Store in a dry, cool and well-ventilated place.
  • Keep container closed when not in use.
  • Keep/Store away from direct sunlight, extremely high or low temperatures and incompatible materials.
  • WHMIS Classification: Class D Division 2 Subdivision B - Toxic material causing other toxic effects
TRUSTED_SUSTAINABILITY

Specifications

Quantity 2 mL
CAS 9002-93-1
Content And Storage Store the unopened product at 2°C to 8°C
Product Line R&D Systems
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.