Learn More
Thermo Scientific™ T4 DNA Ligase Buffer (10X)
Product Code. 11889622 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1.5 mL
Unit Size:
Pack of 10
Product Code. Quantity Unit Size
11889622 1.5 mL Pack of 10
1 options
Product Code. 11889622 Supplier Thermo Scientific™ Supplier No. B69
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
Safely ligate samples with this buffer, for use with T4 DNA Ligase and tested for the absence of endo- and exodeoxyribonucleases and ribonucleases.
10X T4 DNA Ligase buffer is used with T4 DNA Ligase.
TRUSTED_SUSTAINABILITY
Specifications
Concentration 10X
Quantity 1.5 mL
Product Type T4 DNA Ligase Buffer