missing translation for 'onlineSavingsMsg'
Learn More

11-Aminoundecanoic Acid, TRC

Product Code. 30335901
Change view
Click to view available options
Quantity:
1 kg
Unit Size:
1kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30335901 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30335901 Supplier Toronto Research Chemicals Supplier No. TRCA6332101KG
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 11-Aminoundecanoic acid
CAS 2432-99-7
Molecular Formula C11 H23 N O2
Quantity 1 kg
Synonym 11-Aminoundecanoic acid, Aminoundecanoic acid, NSC 240503, NSC 662428, Undecanoic acid, 11-amino-, 11-Aminoundecanoic Acid
InChI Formula InChI=1S/C11H23NO2/c12-10-8-6-4-2-1-3-5-7-9-11(13)14/h1-10,12H2,(H,13,14)
SMILES NCCCCCCCCCCC(=O)O
IUPAC Name 11-aminoundecanoic acid
Molecular Weight (g/mol) 201.306
Formula Weight 201.1729
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.