missing translation for 'onlineSavingsMsg'
Learn More

11-Bromo-1-undecene, 90+%

Product Code. 11484897
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11484897 5 g 5g
11494897 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11484897 Supplier Thermo Scientific Alfa Aesar Supplier No. B21849.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 7766-50-9 | C11H21Br | 233.193 g/mol

11-Bromo-1-undecene is used in advanced polymer dielectrics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
11-Bromo-1-undecene is used in advanced polymer dielectrics.

Solubility
Soluble in water, 0.2422 mg/L @ 25°C (est).

Notes
Store in cool, dry place in tightly closed containers. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7766-50-9
Molecular Formula C11H21Br
Molecular Weight (g/mol) 233.193
MDL Number MFCD00040825
InChI Key YPLVPFUSXYSHJD-UHFFFAOYSA-N
Synonym 11-bromo-1-undecene, 10-undecenyl bromide, 1-bromo-10-undecene, undec-10-enyl bromide, 11-bromo-undec-1-ene, 1-undecene, 11-bromo, 11-bromoundecene, undecylenyl bromide, n-undecylenic bromide, w-undecylenyl bromide
PubChem CID 284148
IUPAC Name 11-bromoundec-1-ene
SMILES C=CCCCCCCCCCBr

Especificaciones

Density 1.063
Boiling Point 149°C to 150°C (35 mmHg)
Flash Point >110°C (230°F)
Assay Percent Range 0.96
Refractive Index 1.469
Quantity 5 g
Beilstein 1753231
Solubility Information Soluble in water,0.2422mg/L @ 25°C (est).
Formula Weight 233.2
Percent Purity 96%
Chemical Name or Material 11-Bromo-1-undecene
Mostrar más Mostrar menos

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.