missing translation for 'onlineSavingsMsg'
Learn More

11-Bromo-1-undecene, 90+%

Product Code. 11494897
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11494897 25 g 25g
11484897 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11494897 Supplier Thermo Scientific Alfa Aesar Supplier No. B21849.14
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 7766-50-9 | C11H21Br | 233.193 g/mol

11-Bromo-1-undecene is used in advanced polymer dielectrics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
11-Bromo-1-undecene is used in advanced polymer dielectrics.

Solubility
Soluble in water, 0.2422 mg/L @ 25°C (est).

Notes
Store in cool, dry place in tightly closed containers. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7766-50-9
Molecular Formula C11H21Br
Molecular Weight (g/mol) 233.193
MDL Number MFCD00040825
InChI Key YPLVPFUSXYSHJD-UHFFFAOYSA-N
Synonym 11-bromo-1-undecene, 10-undecenyl bromide, 1-bromo-10-undecene, undec-10-enyl bromide, 11-bromo-undec-1-ene, 1-undecene, 11-bromo, 11-bromoundecene, undecylenyl bromide, n-undecylenic bromide, w-undecylenyl bromide
PubChem CID 284148
IUPAC Name 11-bromoundec-1-ene
SMILES C=CCCCCCCCCCBr

Specifications

Density 1.063
Boiling Point 149°C to 150°C (35 mm)
Flash Point >110°C (230°F)
Assay Percent Range 0.96
Refractive Index 1.469
Quantity 25 g
Beilstein 1753231
Solubility Information Soluble in water,0.2422mg/L @ 25°C (est).
Formula Weight 233.2
Percent Purity 96%
Chemical Name or Material 11-Bromo-1-undecene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.