missing translation for 'onlineSavingsMsg'
Learn More

11-Bromo-1-undecene, TRC

Product Code. 30346388
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30346388 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30346388 Supplier Toronto Research Chemicals Supplier No. TRCB688895500MG
 Restricted Item
Estimated Shipment: 30-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 11-Bromo-1-undecene
CAS 7766-50-9
Molecular Formula C11H21Br
Quantity 500 mg
Synonym 1-Bromo-10-undecene, 10-Undecenyl bromide, NSC 139878
InChI Formula InChI=1S/C11H21Br/c1-2-3-4-5-6-7-8-9-10-11-12/h2H,1,3-11H2
SMILES BrCCCCCCCCCC=C
IUPAC Name 11-bromoundec-1-ene
Molecular Weight (g/mol) 233.19
Formula Weight 232.0827
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.