missing translation for 'onlineSavingsMsg'
Learn More

11-Deoxycorticosterone acetate, 97%, Thermo Scientific Chemicals

Product Code. 15589513 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15589513 5 g 5g
15579513 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15589513 Supplier Thermo Scientific Acros Supplier No. 460470050
Curiosity Goes Beyond
Use promo code 27966
View all promotions
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56-47-3
Molecular Formula C23H32O4
Molecular Weight (g/mol) 372.51
MDL Number MFCD00003660
InChI Key VPGRYOFKCNULNK-ZIUYWEQENA-N
Synonym deoxycorticosterone acetate, desoxycorticosterone acetate, doca, desoxycortone acetate, percotol, cortexone acetate, decosteron, decosterone, sincortex, syncortyl
PubChem CID 5952
ChEBI CHEBI:34671
SMILES [H][C@@]12CC[C@H](C(=O)COC(C)=O)[C@@]1(C)CC[C@@]1([H])[C@@]2([H])CCC2=CC(=O)CC[C@]12C

Specifications

Melting Point 154.0°C to 160.0°C
Loss on Drying 0.5% max.
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Quantity 5 g
IUPAC Name 2-[(1S,3aS,3bS,9aR,9bS,11aS)-9a,11a-dimethyl-7-oxo-1H,2H,3H,3aH,3bH,4H,5H,7H,8H,9H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-1-yl]-2-oxoethyl acetate
Formula Weight 372.5
Percent Purity 97%
Chemical Name or Material 11-Deoxycorticosterone acetate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.