missing translation for 'onlineSavingsMsg'
Learn More

11-Ketotestosterone-[16,16,17-d3], TRC

Product Code. 30389395
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30389395 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30389395 Supplier Toronto Research Chemicals Supplier No. TRCK21000025MG
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 11-Ketotestosterone-[16,16,17-d3]
CAS 2479914-03-7
Molecular Formula C19H23D3O3
Quantity 25 mg
Synonym (17beta)-17-Hydroxyandrost-4-ene-3,11-dione-16,16
InChI Formula InChI=1S/C19H26O3/c1-18-8-7-12(20)9-11(18)3-4-13-14-5-6-16(22)19(14,2)10-15(21)17(13)18/h9,13-14,16-17,22H,3-8,10H2,1-2H3/t13-,14-,16-,17+,18-,19-/m0/s1/i6D2,16D
SMILES O=C1CC[C@@]2(C)C(CC[C@]([C@@](CC([2H])([2H])[C@]3([2H])O)([H])[C@]3(C)C4)([H])[C@]2([H])C4=O)=C1
IUPAC Name (8S,9S,10R,13S,14S,17S)-17-hydroxy-10,13-dimethyl-1,6,7,8,9,10,12,13,14,15,16,17-dodecahydro-3H-cyclopenta[a]phenanthrene-3,11(2H)-dione-16,16,17-d3
Molecular Weight (g/mol) 305.43
Formula Weight 305.21
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.