missing translation for 'onlineSavingsMsg'
Learn More

12-Amino Minocycline (~90%), TRC

Product Code. 30327344
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30327344 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30327344 Supplier Toronto Research Chemicals Supplier No. TRCA6182855MG
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 12-Amino Minocycline
CAS 864073-42-7
Molecular Formula C23 H28 N4 O6
Quantity 5 mg
Synonym 2-Naphthacenecarboxamide, 12-amino-4,7-bis(dimethylamino)-1,4,4a,5,5a,6,11,12a-octahydro-3,10,12a-trihydroxy-1,11-dioxo-, (4S,4aS,5aR,12aS)-, (4S,4aS,5aR,12aS)-12-Amino-4,7-bis(dimethylamino)-1,4,4a,5,5a,6,11,12a-octahydro-3,10,12a-trihydroxy-1,11-dioxo-2-naphthacenecarboxamide, 12-Aminominocycline, Minocycline Hydrochloride Dihydrate Imp. E (EP)
InChI Formula InChI=1S/C23H28N4O6/c1-26(2)12-5-6-13(28)15-10(12)7-9-8-11-17(27(3)4)19(30)16(22(25)32)21(31)23(11,33)20(24)14(9)18(15)29/h5-6,9,11,17,28,30,33H,7-8,24H2,1-4H3,(H2,25,32)/t9-,11-,17-,23-/m0/s1
SMILES CN(C)[C@H]1[C@@H]2C[C@@H]3Cc4c(ccc(O)c4C(=O)C3=C(N)[C@]2(O)C(=O)C(=C1O)C(=O)N)N(C)C
IUPAC Name (4S,4aS,5aR,12aS)-12-amino-4,7-bis(dimethylamino)-3,10,12a-trihydroxy-1,11-dioxo-4a,5,5a,6-tetrahydro-4H-tetracene-2-carboxamide
Molecular Weight (g/mol) 456.49
Formula Weight 456.2009
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.