missing translation for 'onlineSavingsMsg'
Learn More

12-Crown-4, TRC

Product Code. 30361049
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30361049 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30361049 Supplier Toronto Research Chemicals Supplier No. TRCC79001050MG
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 12-Crown-4
CAS 294-93-9
Molecular Formula C8H16O4
Quantity 50 mg
Synonym 12-Crown-4-ether12C4Ethylene oxide cyclic tetramer
InChI Formula InChI=1S/C8H16O4/c1-2-10-5-6-12-8-7-11-4-3-9-1/h1-8H2
SMILES C1COCCOCCOCCO1
IUPAC Name 1,4,7,10-tetraoxacyclododecane
Molecular Weight (g/mol) 176.21
Formula Weight 176.1049
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.