missing translation for 'onlineSavingsMsg'
Learn More

12-Crown-4, TRC

Product Code. 30352285
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Product Code. Quantity unitSize
30352285 100 mg 100mg
1 options
This item is not returnable. View return policy

Product Code. 30352285

Brand: Toronto Research Chemicals TRCC790010100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 12-Crown-4
CAS 294-93-9
Molecular Formula C8H16O4
Quantity 100 mg
Synonym 12-Crown-4-ether12C4Ethylene oxide cyclic tetramer
InChI Formula InChI=1S/C8H16O4/c1-2-10-5-6-12-8-7-11-4-3-9-1/h1-8H2
SMILES C1COCCOCCOCCO1
IUPAC Name 1,4,7,10-tetraoxacyclododecane
Molecular Weight (g/mol) 176.21
Formula Weight 176.1049
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.