missing translation for 'onlineSavingsMsg'
Learn More

12-Hydroxystearic acid, 95%

Product Code. 11420101
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11420101

Brand: Thermo Scientific Alfa Aesar 044810.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 106-14-9 | C18H36O3 | 300.483 g/mol

12-Hydroxystearic acid is used in cosmetics, wax blends, heavy duty greases, polishes, inks and hot melt adhesives, as a lubricant for natural and synthetic rubbers and as a source for industrial oleo chemicals. It is also used as an intermediate in pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 106-14-9
Molecular Formula C18H36O3
Molecular Weight (g/mol) 300.483
MDL Number MFCD00004592
InChI Key ULQISTXYYBZJSJ-UHFFFAOYSA-N
Synonym 12-hydroxystearic acid, octadecanoic acid, 12-hydroxy, harwax a, cerit fac 3, hydrofol acid 200, dl-12-hydroxystearic acid, ceroxin gl, barolub fto, loxiol g 21, stearic acid, 12-hydroxy
PubChem CID 7789
ChEBI CHEBI:85208
IUPAC Name 12-hydroxyoctadecanoic acid
SMILES CCCCCCC(CCCCCCCCCCC(=O)O)O

Specifications

Melting Point 72°C to 76°C
Color White
Linear Formula CH3(CH2)5CH(OH)(CH2)10CO2H
Quantity 5 g
Beilstein 1726730
Solubility Information Soluble in ether,alcohol and chloroform. Insoluble in water.
Formula Weight 300.48
Percent Purity 95%
Physical Form Crystalline
Chemical Name or Material 12-Hydroxystearic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.