missing translation for 'onlineSavingsMsg'
Learn More

12-O-Methyl Clarithromycin (>90%), TRC

Product Code. 30009004
Change view
Click to view available options
Quantity:
1 mg
10 mg
25 mg
Unit Size:
10mg
1mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30009004 1 mg 1mg
30136911 25 mg 25mg
30003570 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30009004 Supplier Toronto Research Chemicals Supplier No. TRCM2946301MG
 Restricted Item
Estimated Shipment: 17-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 128940-83-0
Molecular Formula C39 H71 N O13
Molecular Weight (g/mol) 761.98
Synonym 6,12-Di-O-methylerythromycin, Oxacyclotetradecane, erythromycin deriv., 12-O-Methylclarithromycin, 6,12-Di-O-methylerythromycin A, Clarithromycin Imp. F (EP)
IUPAC Name (3R,4S,5S,6R,7R,9R,11R,12R,13S,14R)-6-[(2S,3R,4S,6R)-4-(dimethylamino)-3-hydroxy-6-methyloxan-2-yl]oxy-14-ethyl-12-hydroxy-4-[(2R,4R,5S,6S)-5-hydroxy-4-methoxy-4,6-dimethyloxan-2-yl]oxy-7,13-dimethoxy-3,5,7,9,11,13-hexamethyl-oxacyclotetradecane-2,10-dione
SMILES CC[C@H]1OC(=O)[C@H](C)[C@@H](O[C@H]2C[C@@](C)(OC)[C@@H](O)[C@H](C)O2)[C@H](C)[C@@H](O[C@@H]3O[C@H](C)C[C@@H]([C@H]3O)N(C)C)[C@@](C)(C[C@@H](C)C(=O)[C@H](C)[C@@H](O)[C@]1(C)OC)OC

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C39H71NO13/c1-16-27-39(10,48-15)32(43)22(4)29(41)20(2)18-38(9,47-14)34(53-36-30(42)26(40(11)12)17-21(3)49-36)23(5)31(24(6)35(45)51-27)52-28-19-37(8,46-13)33(44)25(7)50-28/h20-28,30-34,36,42-44H,16-19H2,1-15H3/t20-,21-,22+,23+,24-,25+,26+,27-,28+,30-,31+,32-,33+,34-,36+,37-,38-,39-/m1/s1
Formula Weight 761.4925
Percent Purity >95%
Chemical Name or Material 12-O-methyl Clarithromycin
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.