missing translation for 'onlineSavingsMsg'
Learn More

LMS™ Series 4 Model 1200NP Cooled Incubator, 1200 L, Stainless Steel

Product Code. 11897602
Click to view available options
Model:
1200NP
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 11897602

Brand: LMS™ 1200NP

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Temperature Range: -10°C to +50°C

  • Reliable and accurate temperature control
  • Temperature range -10.0°C to +50.0°C
  • Models within this series include 600L and 1200L capacity incubators as either single or dual temperature (24 hour or 7 day cycle) versions, or a fully programmable 1200L capacity incubator providing multiple temperature control
  • Stainless steel exterior and interior construction with foamed polyurethane insulation
  • Energy efficient and eco-friendly
  • P.I.D. heating and cooling to ensure stable internal temperature
  • Pre-set safety cut-out
  • Hermetically sealed refrigeration system
  • Door lock
  • Controls easy to read and simple to use
  • Temperature variation with average workload ±0.5°C
  • Full range of factory-fitted optional extras available, including LED lighting, access porthole, inner perspex doors, outer glazed doors, alarm system, internal socket and chart recorder
  • Ideal for applications such as growth studies, B.O.D.tests, seed germination and vernalisation, tissue culture, enzymology, serum studies and immunology, sample storage or product shelf life tests

Specifications

Certifications/Compliance ISO 9001:2008
Controller Type Programmable
Description Freestanding incubator
Capacity (Metric) 1200 L
Chamber Material Stainless Steel
Convection Type Fan-assisted Circulation
Dimensions (D x W) Shelf 550 mm x 530 mm
Lighting Optional LED
Material Stainless Steel
No. of Shelves 10
Temperature Range (Metric) -10°C to +50°C
Type Refrigerated Incubator
Display Digital
Insulation Foamed polyurethane
Model 1200NP
No. of Doors 2
Refrigerant CFC-free
Temperature Stability ±0.1°C
Temperature Uniformity ±0.5°C
Warranty 2 Years
Width (English) 59.0 in.
Width (English) Interior 53.1 in.
Width (Metric) 1500 mm
Width (Metric) Interior 1350 mm
Height (English) 81.9 in.
Height (English) Interior 59.0 in.
Height (Metric) 2080 mm
Height (Metric) Interior 1500 mm
Depth (English) Interior 26.4 in.
Depth (Metric) Interior 670 mm
Weight (Metric) 235 kg
Electrical Requirements 240 V, 50 Hz
Voltage 230 V
Frequency 50 Hz
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.