Learn More
Epredia™ PrintMate™ AS Cassette Printer Accessories
Product Code. 12648206 Shop All Epredia Products
Change view
Click to view available options
Product Type:
Cassette Hopper
Collection System
Collection Tray
Computer
Hot Foil Tape
Unit Size:
Each
Pack of 4
Pack of 5
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type unitSize
12648206 Cassette Hopper Each
12973745 Collection System Each
15589535 Collection Tray Pack of 4
12628206 Hot Foil Tape Pack of 5
12963815 Computer Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 12648206 Supplier Epredia™ Supplier No. A79510024
Only null left
Add to basket
Add to basket
Maintain the Epredia™ PrintMate™ Cassette Printer with PrintMate™ Accessories.
Accessories Include:
  • Collection system collection trays (box of 4)
  • Hopper
  • Hot foil tape, box of 5 (roll of tape prints approx. 20,000 cassettes)
TRUSTED_SUSTAINABILITY
Specifications
Product Type Cassette Hopper
Description Cassette Hopper (each)
For Use With (Equipment) PrintMate AS Cassette Printer, PrintMate Cassette Printer