Learn More
Sartorius™ Cellulose Nitrate (CN) Membrane Filters
Ideal membrane filters for colony counting, particle testing and microscopy. Sartorius™ Cellulose Nitrate (CN) Membrane Filters provide effective retention, high flow rates and optimal colony growth.
Brand: Sartorius™ 13004--47------N
Code : NEW
Additional Details : Weight : 0.01000kg Transport : UN number : 3270 Chem class : 4.1 Pack group : II
Description
- Gridded
- Non-sterile
- Hydrophilic
- Cellulose nitrate (CN; cellulose mixed water)
- Market:
- Environmental/water testing
- Food and beverage
- Application:
- Microbial enumeration
Specifications
Gray/White | |
Cellulose Nitrate | |
0.8 μm | |
Used for colony counting, particle testing and microscopy | |
Membrane Filter |
Membrane Filter | |
Cellulose Nitrate | |
Non-sterile | |
100 Pack | |
47 mm |
Safety and Handling
- X100 MEMBRANE NC G/W 0,8µ 47MM D=47mm, grau/weiß, (VE=100 Stck.)
Signal Word
- Danger
Hazard Category
- Flammable solid Category 2
Hazard Statement
- H225-Highly flammable liquid and vapour.
Precautionary Statement
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
- MIXTURE LIST-Contains : nitrocellulose