Learn More
Sartorius™ 13107 Series Gridded Sterile Cellulose Nitrate (CN) Membrane Filters
Brand: Sartorius™ 13107--47----HCN
Code : NEW
Additional Details : Weight : 0.01000kg Transport : UN number : 3270 Chem class : 4.1 Pack group : II
Specifications
White | |
Cellulose Nitrate | |
0.2 μm | |
Used for colony counting, particle testing and microscopy | |
100 Pack | |
Membrane Filter | |
47 mm |
Membrane Filter | |
Cellulose Nitrate | |
Non-sterile | |
Black | |
115 to 145 μm | |
47 mm |