missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ 13mm PP Screw Cap, 13-425 thread

Product Code. 11797597 Shop All Fisherbrand Products
Change view
Click to view available options
Color:
Black
White
Product Type:
Center Hole Screw Cap
Closed Top Screw Cap
Unit Size:
Pack of 1000
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Product Type unitSize
11797597 Black Center Hole Screw Cap Pack of 1000
11707607 Black Closed Top Screw Cap Pack of 1000
11717607 White Center Hole Screw Cap Pack of 1000
11898881 White Closed Top Screw Cap Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 11797597
Sweet Deals on Top Products
Use promo code 28057
View all promotions
Estimated Shipment: 14-08-2026
Add to basket
Add to basket

Polypropylene caps are chemically inert and suitable for most chromatography applications.

  • 13mm
  • PP Screw Cap
  • Center hole
TRUSTED_SUSTAINABILITY

Specifications

Color Black
Diameter (Metric) Outer 13 mm
Material Polypropylene
Quantity 100
Closure Size ND13
Product Type Center Hole Screw Cap
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.