missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ 13mm PP Screw Seal, Black, Closed Top, 13-425 Thread, Assembled Septum

Product Code. 11884970 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Color:
Beige, Red-Orange, Black
Black, Red-Orange, Transparent
Cream, Black
Gray, Red, Black
Material (Liner):
Natural Rubber/TEF
PTFE/Butyl
Red Rubber/PTFE
Silicone/PTFE
Thickness (Metric):
1 mm
1.3 mm
1.5 mm
Unit Size:
Pack of 1000
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Material (Liner) Thickness (Metric) Unit Size
11884970 Cream, Black Silicone/PTFE 1.5 mm Pack of 1000
11823861 Gray, Red, Black PTFE/Butyl 1.3 mm Pack of 1000
11864970 Black, Red-Orange, Transparent Natural Rubber/TEF 1.3 mm Pack of 1000
12636715 Beige, Red-Orange, Black Red Rubber/PTFE 1 mm Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 11884970
Spotlight Offers
Use promo code 28038
View all promotions
Estimated Shipment: 04-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

Polypropylene caps are chemically inert and suitable for most chromatography applications.

  • 13mm
  • PP
  • Screw Cap
  • Black
  • Pre-assembled caps and septa are convenient and minimize contamination from handling
TRUSTED_SUSTAINABILITY

Specifications

Material (Liner) Silicone/PTFE
Color Cream, Black
Diameter (Metric) Outer 13 mm
Material Polypropylene
For Use With (Application) HPLC
Quantity 100
Thickness (Metric) 1.5 mm
Product Type Screw Cap
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.