missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Plastic Lock Bacti Caps

Product Code. 11537143 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Color:
Black
Blue
Red
White
Yellow
Unit Size:
Pack of 100
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
11537143 Yellow Pack of 100
11547143 Blue Pack of 100
11527143 Red Pack of 100
11507143 Black Pack of 100
11517143 White Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 11537143
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

Made from chemically resistant virgin plastic, Bacti-Caps™ provide a firm, positive culture tube closure allowing a controlled gas interchange. They are ideal for use in science, research, and other laboratories.

  • Bacti-Caps™  for test tubes without edge
  • Provide a firm, positive culture tube closure allowing a controlled gas interchange, thus proving a more practical alternative to cotton wool plugs
  • Suitable for all standard test tube sizes, they fit rimless and plastic tubes with an outside diameter of 13mm, 16mm, 19mm, 25mm and 38mm 
  • The three flexible fins molded on the inner wall of each cap ensure a positive fit even when minor variations in tube size occur
  • Bacti-Caps™ are made from chemically resistant virgin plastic so they are unaffected by culture media, most acids, alkalis, alcohols and esters
  • Improved culture growth
  • Color coded
  • Controlled gas interchange

Specifications

Product Type Lock Bacti Cap
Autoclavable Autoclavable
Material Plastic
Color Yellow
Quantity 100 Pack
Size (Metric) 13 mm
For Use With (Equipment) Test Tubes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.