Learn More
Hamilton™ 14 gauge, Hubless Needle, custom length (0.375 to 12 in), point style 5 or AS, 6/PK
Click to view available options
Needle Gauge:
14 G
Unit Size:
Pack of 6
Description
Specification Sheet
- Gauge – 14 gauge
- Point Style – 5 or AS
- Needle Length – Custom Needle Length
- Autoclavable – Yes
- Sales Quantity – 6 needles per pack
- Hub – No Hub
Specifications
Specifications
| Needle Gauge | 14 G |
| Product Type | Hubless Needles |