missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Labcon™ ViewPoint™ 15 mL Polypropylene Thermochromic Centrifuge Tubes

Click to view available options
No. per Pack:
10/IntegraPack™ Bag
25/reusable polypropylene rack
Sterility:
Non-sterile
Sterile
Unit Size:
Pack of 50
Description
- Real-time temperature signaling
- Reversibly transition from clear to light blue at 4°C with full dark blue transition at 0°C
- With attached flat screw caps
- Black graduations and white writing surface
- ATP, RNase/DNase and Endotoxin Free (non-pyrogenic)
- Sperm Motility Assay (SMA) Mouse Embryo Assay (MEA) validated safe for cell culture
- Polypropylene USP Class VI resin
- Centrifuging to 12,500 x G
- Freezable to -80°C
- Made in USA
Specifications
Specifications
| Certifications/Compliance | ISO 9001:2015; FDA CFR21 GMP |
| Closure Material | Polypropylene |
| Closure Color | Purple |
| Color | Clear to Blue color change at 0°C to 4°C |
| Capacity (Metric) | 15 mL |
| Disposable | Single-use |
| Material | Polypropylene |
| For Use With | Centrifuges, Rotators, Shakers |
| No. per Pack | 25/reusable polypropylene rack |
| Packaging | 25 Tubes/Reusable Rack, 2 Racks/Pk., 6 Pk./Cs. |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction