missing translation for 'onlineSavingsMsg'
Learn More

TdB Labs AB 150 kDa Q-dextran for Ion-exchange Chromatography and Biomolecular Studies

Product Code. 30216348
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30216348 1 g 1g
30216320 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30216348 Supplier TdB Labs AB Supplier No. QD150_1g
 Restricted Item
Estimated Shipment: 05-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Q-dextran is a positively charged dextran modified with quaternary ammonium groups. It is used in ion exchange chromatography, biochemical assays, and for studying electrostatic interactions with negatively charged biomolecules.

  • Storage and Stability:
    • Store dry at room temperature, protected from moisture and light.
    • Stable for extended periods under recommended storage conditions without loss of charge properties.
  • Specifications

    Quantity 1 g
    Product Type Modified Dextran
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.