missing translation for 'onlineSavingsMsg'
Learn More

(15S)-Latanoprost, TRC

Product Code. 30040388
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30040388 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30040388 Supplier Toronto Research Chemicals Supplier No. TRCL17728550MG
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 145773-22-4
Molecular Formula C26 H40 O5
Quantity 50 mg
Synonym Isopropyl (Z)-7-[(1R,2R,3R,5S)-3,5-Dihydroxy-2-[(3S)-3-hydroxy-5-phenylpentyl]cyclopentyl]-5-heptenoate, (15S)-Latanoprost, Latanoprost Related Compound B (USP)
InChI Formula InChI=1S/C26H40O5/c1-19(2)31-26(30)13-9-4-3-8-12-22-23(25(29)18-24(22)28)17-16-21(27)15-14-20-10-6-5-7-11-20/h3,5-8,10-11,19,21-25,27-29H,4,9,12-18H2,1-2H3/b8-3-/t21-,22-,23-,24+,25-/m1/s1
SMILES CC(C)OC(=O)CCC\C=C/C[C@H]1[C@@H](O)C[C@@H](O)[C@@H]1CC[C@H](O)CCc2ccccc2
IUPAC Name propan-2-yl (Z)-7-[(1R,2R,3R,5S)-3,5-dihydroxy-2-[(3S)-3-hydroxy-5-phenylpentyl]cyclopentyl]hept-5-enoate
Molecular Weight (g/mol) 432.59
Formula Weight 432.2876
Chemical Name or Material Isopropyl (Z)-7-[(1R,2R,3R,5S)-3,5-dihydroxy-2-[(3S)-3-hydroxy-5-phenylpentyl]cyclopentyl]-5-heptenoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.