Description
The Thermo Scientific™ AI/AS 1610 Autosampler elevates user experience on liquid sampling automation for GC and GC-MS through simplicity, robustness, and throughput capability.
Enhance liquid sampling automation for your gas chromatography (GC) applications with simplicity and robustness. Designed to meet the demands of any busy high-throughput lab, the Thermo Scientific™ AI/AS 1610 Liquid Autosampler will help to process more samples efficiently and safely with its advanced design and usability features, greatly enhanced for easy adoption for both beginners and experienced users. With the possibility to expand to up to a 310-sample capacity with simultaneous dual-tower operations, the AI/AS 1610 offers the fit-for-purpose solution to budget-conscious laboratories looking for reliable automation of liquid samples injection into a GC or GC-MS system.
Combine performance, simplicity, and robustness for your every-day workflow.
The AI/AS 1610 Liquid Autosampler is available in two models: the upgradable 8-vial capacity AI 1610 and the 155-vial capacity AS 1610. Both offers the dual-tower Gemini configuration for simultaneous injection in two channels.
Enhanced usability
- Easy set up through self-aligning not requiring any position adjustment
- Easy slide-in installation and free access to the injector for maintenance
- Easy syringe viewing and replacement with illuminated compartment and magnifier lens
- Enhanced color-coded instrument status indication
- Safe operation with automatic selection of the best injection settings through recognition of the injector type and injection mode
- Simplified connectivity with a single cable including communication, synchronization, and power supply
Enhanced sample throughput
- Wide sample capacity scalability to fit any workload demand
- Dual Tower Gemini for upmost productivity with simplified configuration and connectivity
Enhanced performance
- Expanded applicability with liquid sample volume injection up to 80μL
- Free pre- and post-injection syringe rinsing with single or combined solvents (up to four) virtually eliminates sample carry-over
- Efficient, accurate sampling of low boiling compounds with the syringe kept away from the inlet’s temperature influence
- Seamless integration into Thermo Scientific™ chromatography and™ software
Spécification
Spécification
| Sample Capacity | 155 x 2 mL Vials |
| Injection Mode | Automatically selected and optimized according to injector type |
| Injection Speed | Fast/Slow/Variable (1 to 60,000.00 μL/min.) |
| Injection Volume | 0.01 to 80 μL |
| Description | AS 1610-155 Liquid Autosampler |
| For Use With (Equipment) | TRACE 1600 Series |
| Washing Solvents | Up to 4 |
| Quantity | Each |
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.