missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ 17-AAG
GREENER_CHOICE

Product Code. 18004802
missing translation for 'orderingAttributeHoverText'
Quantity:
1 mg
missing translation for 'unitSize'
1mg
This item is not returnable. View return policy

Product Code. 18004802

missing translation for 'mfr': Tocris Bioscience™ 1515/1

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Selective Hsp90 inhibitor

Inhibitor of heat shock protein 90 (Hsp90) chaperone activity, and an analog of geldanamycin (Cat. No. 1368). Subsequently inhibits the activity of oncogenic proteins such as p185erbB-2 (IC50 = 31 nM), N-ras, Ki-ras and c-Akt. Antitumor in vivo. Also protects neuroprogenitor cells against stress-induced apoptosis at low concentrations (10 nM) in vitro.
TRUSTED_SUSTAINABILITY

Specifications

CAS 75747-14-7
Quantity 1 mg
Molecular Formula C31H43N3O8
Formula Weight 585.7
Purity >98%
Molecular Weight (g/mol) 585.698
Chemical Name or Material 17-AAG
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.