missing translation for 'onlineSavingsMsg'
Learn More

17α-Acetyloxy-11β-methyl-19-norprogesterone, TRC

Product Code. 30414486
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30414486 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30414486 Supplier Toronto Research Chemicals Supplier No. TRCN6865005MG
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Norgestomet
CAS 25092-41-5
Purity Grade Notes HPLC
Molecular Formula C23 H32 O4
Quantity 5 mg
Synonym 17alphalpha-Acetyloxy-11beta-methyl-19-norprogesterone, 19-Norpregn-4-ene-3,20-dione','17-hydroxy-11β-methyl-','acetate (8CI), (11β)-17-(Acetyloxy)-11-methyl-19-norpregn-4-ene-3,20-dione, 17α-Acetoxy-11β-methyl-19-norpregn-4-ene-3,20-dione, Crestar, Norgestomet, Norprogesterone, SC 21009
InChI Formula InChI=1S/C23H32O4/c1-13-12-22(4)20(9-10-23(22,14(2)24)27-15(3)25)19-7-5-16-11-17(26)6-8-18(16)21(13)19/h11,13,18-21H,5-10,12H2,1-4H3/t13-,18-,19-,20-,21+,22-,23-/m0/s1
SMILES C[C@H]1C[C@@]2(C)[C@@H](CC[C@]2(OC(=O)C)C(=O)C)[C@@H]3CCC4=CC(=O)CC[C@@H]4[C@@H]13
IUPAC Name [(8R,9S,10R,11S,13S,14S,17R)-17-acetyl-11,13-dimethyl-3-oxo-1,2,6,7,8,9,10,11,12,14,15,16-dodecahydrocyclopenta[a]phenanthren-17-yl] acetate
Molecular Weight (g/mol) 372.498
Formula Weight 372.23
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.