missing translation for 'onlineSavingsMsg'
Learn More

17Beta-Acetyl-estradiol 3-(Tri-O-acetyl-Beta-D-glucuronic Acid Methyl Ester), TRC

Product Code. 30318881
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30318881 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30318881 Supplier Toronto Research Chemicals Supplier No. TRCA173825100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 17beta-Acetyl-estradiol 3-(Tri-O-acetyl-beta-D-glucuronic Acid Methyl Ester)
CAS 59495-70-4
Molecular Formula C33H42O12
Quantity 100 mg
Synonym (17beta)-17-Acetoxyestra-1,3,5(10)-trien-3-yl Tri-O-acetyl-beta-D-glucuronic Acid Methyl Ester, 17-Acetyl-estradiol Tri-O-acetyl-beta-D-glucuronic Acid Methyl Ester
InChI Formula InChI=1S/C33H42O12/c1-16(34)40-26-12-11-25-24-9-7-20-15-21(8-10-22(20)23(24)13-14-33(25,26)5)44-32-30(43-19(4)37)28(42-18(3)36)27(41-17(2)35)29(45-32)31(38)39-6/h8,10,15,23-30,32H,7,9,11-14H2,1-6H3/t23-,24-,25+,26+,27+,28+,29+,30-,32-,33+/m1/s1
SMILES COC(=O)[C@H]1O[C@@H](Oc2ccc3[C@H]4CC[C@]5(C)[C@H](CC[C@H]5[C@@H]4CCc3c2)OC(=O)C)[C@H](OC(=O)C)[C@@H](OC(=O)C)[C@@H]1OC(=O)C
IUPAC Name methyl (2S,3S,4S,5R,6S)-3,4,5-triacetyloxy-6-[[(8R,9S,13S,14S,17S)-17-acetyloxy-13-methyl-6,7,8,9,11,12,14,15,16,17-decahydrocyclopenta[a]phenanthren-3-yl]oxy]oxane-2-carboxylate
Molecular Weight (g/mol) 630.68
Formula Weight 630.2676
Product Title
Select an issue

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.